Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1530 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562252
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562252 Supplier Invitrogen™ Supplier No. PA562252
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQEAQKLAAE RARAPVVPYG VDLHYWGQEL PTAGKIVKSD SQHRFWKPSE VEEEVVNADI SEMLRSRHIT FAGKFEPVQH WCRAPRPDGR LCERQDRLK Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

The protein encoded by KIAA1530 appears to be involved in ubiquitination and dephosphorylation of RNA polymerase II subunits that stall after UV irradiation. The encoded protein interacts with several members of the nucleotide excision repair complex, and is thought to be involved in the transcription-coupled nucleotide excision repair (TC-NER) pathway to help remove lesions in the DNA that block transcription. Defects in KIAA1530 can cause UV-sensitive syndrome 3. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1530
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UVSSA
Gene Accession No. Q2YD98
Gene Alias KIAA1530; UV stimulated scaffold protein A; UVSS3; UVSSA; UV-stimulated scaffold protein A
Gene Symbols UVSSA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1530. Recombinant protein control fragment (Product #RP-100208).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57654
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.