Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIAA1530 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 250541AP150UL
Description
The protein encoded by KIAA1530 appears to be involved in ubiquitination and dephosphorylation of RNA polymerase II subunits that stall after UV irradiation. The encoded protein interacts with several members of the nucleotide excision repair complex, and is thought to be involved in the transcription-coupled nucleotide excision repair (TC-NER) pathway to help remove lesions in the DNA that block transcription. Defects in KIAA1530 can cause UV-sensitive syndrome 3. Alternative splicing results in multiple transcript variants.Specifications
| KIAA1530 | |
| Polyclonal | |
| Unconjugated | |
| UVSSA | |
| KIAA1530 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57654 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q2YD98 | |
| UVSSA | |
| KIAA1530 Fusion Protein Ag21868 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction