Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1551 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553344
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553344 Supplier Invitrogen™ Supplier No. PA553344
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEALSNKASK KICVKNVPCD SEHMRPSKLA VQVESCGKSN EKHSSGVQTS KESLNGLTSH GKNLKIHHSQ ESKTYNILRN VKEKVGGKQP DKIWIDKTKL DKLTNISNEA QFSQMPPQVK DQKKLYLNRV GFKCTERE Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 49%.

Plays a role in the regulation of imprinted gene expression, regulates repressive epigenetic modifications associated with SETDB1. Required for the recruitment or accumulation of SETDB1 to the endogenous retroviruses (ERVs) and maintenance of repressive chromatin configuration, contributing to a subset of the SETDB1-dependent ERV silencing in embryonic stem cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1551
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA1551
Gene Accession No. Q9HCM1
Gene Alias C12orf35; KIAA1551; RESF1; Retroelement silencing factor 1; uncharacterized protein C12orf35; uncharacterized protein KIAA1551; UTA2-1
Gene Symbols KIAA1551
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1551. Recombinant protein control fragment (Product #RP-90772).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55196
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.