Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1551 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546195 Supplier Invitrogen™ Supplier No. PA546195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KEQAPLQVSG IKSTKEDWLK FVATKKRTQK DSQERDNVNS RLSKRSFSAD Sequence homology: Human: 100%.

Plays a role in the regulation of imprinted gene expression, regulates repressive epigenetic modifications associated with SETDB1. Required for the recruitment or accumulation of SETDB1 to the endogenous retroviruses (ERVs) and maintenance of repressive chromatin configuration, contributing to a subset of the SETDB1-dependent ERV silencing in embryonic stem cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1551
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KIAA1551
Gene Accession No. Q9HCM1
Gene Alias C12orf35; KIAA1551; RESF1; Retroelement silencing factor 1; uncharacterized protein C12orf35; uncharacterized protein KIAA1551; UTA2-1
Gene Symbols KIAA1551
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human KIAA1551 (aa 1661-1710).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55196
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.