Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1586 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557630 Supplier Invitrogen™ Supplier No. PA557630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NKTTRQASLR KKIREHDVSK AHGKIQDLLK ESTNDSICNL VHKQNNKNID ATVKVFNTVY SLVKHNRPLS DIEGARELQE KNGEVNCLNT RYSATRI Highest antigen sequence identity to the following orthologs: Mouse - 24%, Rat - 24%.

E3 SUMO-protein ligase; facilitates UBE2I/UBC9-mediated SUMO2 modification of target proteins (PubMed:26524493).
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1586
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA1586
Gene Accession No. Q9HCI6
Gene Alias E3 SUMO-protein ligase KIAA1586; E3 SUMO-protein transferase KIAA1586; KIAA1586
Gene Symbols KIAA1586
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1586. Recombinant protein control fragment (Product #RP-94942).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57691
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.