Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA1609 Monoclonal Antibody (OTI3C4), TrueMAB™, OriGene
SDP

Catalog No. 502542610
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2610 30 μL
50-167-2151 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2610 Supplier OriGene Supplier No. TA501008S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KIAA1609 Monoclonal antibody specifically detects KIAA1609 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen KIAA1609
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3C4
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Srr
Gene Accession No. Q9GZT4
Gene Alias 4632415K11Rik; Kiaa1609; MEAK7; MTOR associated protein, eak-7 homolog; MTOR-associated protein MEAK7; TBC/LysM associated domain containing 1; TBC/LysM-associated domain containing 1; TBC/LysM-associated domain-containing protein 1; TLD domain containing 1; TLD domain-containing protein 1; TLDC1
Host Species Mouse
Immunogen Full length human recombinant protein of human SRR produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63826
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.