Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1671 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553737 Supplier Invitrogen™ Supplier No. PA553737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PISHSLRRSR FSESESRSPL EDETDNTWMF KDSTEEKSPR KEESDEEETA SKAERTPVSH PQRMPAFPGM DPAVLKAQLH KRPEVDSPGE TPSWAPQPKS PKSPFQPGVL GSRVLPSSMD KDERSDEPSP QWLKELKSKK RQSL Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1671
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.80 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA1671
Gene Accession No. Q9BY89
Gene Alias CTA-221G9.5; KIAA1671; Uncharacterized protein KIAA1671
Gene Symbols KIAA1671
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1671. Recombinant protein control fragment (Product #RP-91376).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85379
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.