Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1680 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559770 Supplier Invitrogen™ Supplier No. PA559770
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEHSTNKNVF INCLSSGKSE GDDSGFTEDQ TRRSVKQSTR KLLPKSFSSH YKFSKPVLQS QSISLVQQSE FSLEVTQYQE REPVLVRASP SCSVDVTERA Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1680
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCSER1
Gene Accession No. Q9C0I3
Gene Alias CCSER1; coiled-coil serine rich protein 1; coiled-coil serine-rich protein 1; FAM190A; family with sequence similarity 190, member A; KIAA1680; protein FAM190A; serine-rich coiled-coil domain-containing protein 1
Gene Symbols CCSER1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1680. Recombinant protein control fragment (Product #RP-100265).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401145
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.