Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1683 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546503
Description
Peptide sequence: LPDKMEKAPP QPQHEGLKSK EHLPQQPAEG KTASRRVPRL RAVVESQAFK Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
| KIAA1683 | |
| Polyclonal | |
| Unconjugated | |
| KIAA1683 | |
| IQ domain-containing protein N; IQCN; KIAA1683; uncharacterized protein KIAA1683 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80726 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H0B3 | |
| KIAA1683 | |
| Synthetic peptide directed towards the N-terminal of human KIAA1683. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction