Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1688 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572692
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572692 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572692 Supplier Invitrogen™ Supplier No. PA572692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIAA1688 Polyclonal Antibody for Western Blot, ELISA

Members of the F-box protein family, such as FBXO28, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1688
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ARHGAP39
Gene Accession No. P18890, P59281, Q9C0H5
Gene Alias 9530053N22; AI843066; ARHGAP39; CrGAP; crossGAP homolog; D15Wsu169e; KIAA1688; Porf-2; preoptic regulatory factor 2; preoptic regulatory factor-2; Rho GTPase activating protein 39; rho GTPase-activating protein 39; RhoGAP93B homolog; Vilse
Gene Symbols ARHGAP39
Host Species Rabbit
Immunogen ARHGAP39 antibody for an 18 amino acid synthetic peptide near the carboxy terminus of human ARHGAP39. The immunogen is within amino acids 880 - 930 of ARHGAP39.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223666, 500901, 80728
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.