Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1731NL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555246
Description
Immunogen sequence: CSGWSSSVIW RHTQFAVERC GFCGSSGPGA PLEPSTLGSK HLPWEAVSAG FADRNRNMDG AMWLSLCPDN EDLLWRKKHK LLQAR Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 25%.
Specifications
| KIAA1731NL | |
| Polyclonal | |
| Unconjugated | |
| CEP295NL | |
| CEP295 N-terminal like; CEP295 N-terminal-like protein; CEP295NL; DDC8; differential display clone 8; KIAA1731 N-terminal like protein; KIAA1731NL; protein DDC8 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100653515 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96MC4 | |
| CEP295NL | |
| Recombinant protein corresponding to Human KIAA1731NL. Recombinant protein control fragment (Product #RP-93462). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction