Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIAA1731NL Polyclonal Antibody

KIAA1731NL Polyclonal Antibody
Supplier: Thermo Scientific PA554842
Description
KIAA1731NL Polyclonal Antibody for IHC
Specifications
| KIAA1731NL | |
| Polyclonal | |
| Unconjugated | |
| CEP295NL | |
| CEP295 N-terminal like; CEP295 N-terminal-like protein; CEP295NL; DDC8; differential display clone 8; KIAA1731 N-terminal like protein; KIAA1731NL; protein DDC8 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100653515 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96MC4 | |
| CEP295NL | |
| Recombinant protein corresponding to Human KIAA1731NL. Recombinant protein control fragment (Product #RP-93461). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction