Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1751 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555889
Description
Immunogen sequence: HLRDFIHVDF DPPGPLSAGM SCEVLVTFKP MINKDLEGNI SFLAQTGEFS VPLKCSTKKC SLSLDKELID FGSYVVGETT SRTITLTN Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.
As part of the central apparatus of the cilium axoneme may play a role in cilium movement. [UniProt].
Specifications
| KIAA1751 | |
| Polyclonal | |
| Unconjugated | |
| CFAP74 | |
| C1orf222; CFAP74; cilia and flagella associated protein 74; cilia- and flagella-associated protein 74; FLJ45476; hCG1647286-like; KIAA1751; LOW QUALITY PROTEIN: cilia- and flagella-associated protein 74 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 85452 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C0B2 | |
| CFAP74 | |
| Recombinant protein corresponding to Human KIAA1751. Recombinant protein control fragment (Product #RP-94252). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction