Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1751 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556101 Supplier Invitrogen™ Supplier No. PA556101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDVPVESLTA IPVFDPRHRE ASSRPGPLSP EAEELRPILV TLDYIQFDTD TPAPPATREL QVGCIRTTQP SPKK Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 69%.

As part of the central apparatus of the cilium axoneme may play a role in cilium movement. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1751
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP74
Gene Accession No. Q9C0B2
Gene Alias C1orf222; CFAP74; cilia and flagella associated protein 74; cilia- and flagella-associated protein 74; FLJ45476; hCG1647286-like; KIAA1751; LOW QUALITY PROTEIN: cilia- and flagella-associated protein 74
Gene Symbols CFAP74
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1751. Recombinant protein control fragment (Product #RP-94581).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85452
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.