Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1751 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571023
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 100%; Human: 100%; Rat: 90%.
As part of the central apparatus of the cilium axoneme may play a role in cilium movement. [UniProt].
Specifications
KIAA1751 | |
Polyclonal | |
Unconjugated | |
CFAP74 | |
C1orf222; CFAP74; cilia and flagella associated protein 74; cilia- and flagella-associated protein 74; FLJ45476; hCG1647286-like; KIAA1751; LOW QUALITY PROTEIN: cilia- and flagella-associated protein 74 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
85452 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9C0B2 | |
CFAP74 | |
synthetic peptide directed towards the C-terminal region of human C1orf222. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction