Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1751 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546504
Description
Peptide sequence: AATSFSEQQL EGTESSQADM QSRKELEKLD KEQEEEQPAG EPGCQAKPGW Sequence homology: Human: 100%.
As part of the central apparatus of the cilium axoneme may play a role in cilium movement. [UniProt].
Specifications
| KIAA1751 | |
| Polyclonal | |
| Unconjugated | |
| CFAP74 | |
| C1orf222; CFAP74; cilia and flagella associated protein 74; cilia- and flagella-associated protein 74; FLJ45476; hCG1647286-like; KIAA1751; LOW QUALITY PROTEIN: cilia- and flagella-associated protein 74 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 85452 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9C0B2 | |
| CFAP74 | |
| Synthetic peptide directed towards the C-terminal of human KIAA1751. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction