Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIAA1958 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | KIAA1958 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
KIAA1958 Polyclonal specifically detects KIAA1958 in Human samples. It is validated for Western Blot.Specifications
| KIAA1958 | |
| Polyclonal | |
| Rabbit | |
| FLJ39294, hypothetical protein LOC158405, KIAA1958, MGC142075 | |
| KIAA1958 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 158405 | |
| Synthetic peptides corresponding to KIAA1958(KIAA1958) The peptide sequence was selected from the C terminal of KIAA1958. Peptide sequence SPITLLSTVVKYNSQYLNMRTLQEHADLMYGDIELLKDPQNQPYFARTDS. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title