Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1958 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569850
Description
This target displays homology in the following species: Cow: 85%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
KIAA1958 is a protein coding gene.
Specifications
KIAA1958 | |
Polyclonal | |
Unconjugated | |
KIAA1958 | |
KIAA1958; KIAA1958 ortholog; uncharacterized protein KIAA1958; uncharacterized protein KIAA1958 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
158405 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N8K9 | |
KIAA1958 | |
synthetic peptide directed towards the C terminal region of human KIAA1958. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction