Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA1967 Mouse anti-Human, Monkey, Mouse, Rat, Clone:3G4-D11-D7, Abnova™

Catalog No. 89333886 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-886 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-886 Supplier Abnova Corporation Supplier No. MAB17664

Mouse monoclonal antibody raised against recombinant human KIAA1967.

Specifications

Antigen KIAA1967
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3G4-D11-D7
Conjugate Unconjugated
Description KIAA1967
Dilution Immunoprecipitation
Immunocytochemistry (1:200)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.1M Tris-Glycine, 150 mM NaCl, pH 7.4 (50% glycerol, 0.2% sodium azide)
Gene KIAA1967
Gene Alias DBC-1, DBC1
Host Species Mouse
Immunogen Recombinant protein corresponding to a fragment of human KIAA1967 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 57805
Test Specificity This antibody detects endogenous levels of KIAA1967 and does not cross-react with related proteins.
Target Species Human, Monkey, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.