Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA2013 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566335
Description
Immunogen sequence: EMLELLDMPAA ELLQDHQLLW AQLFSPGVEM KKITDTHTPS GLTVNLTLYY MLSCSPAPLL SPSLSHRERD QMESTLNYED HC Highest antigen sequence identity to the following orthologs - mouse 92%, rat 93%.
KIAA2013 is a protein coding gene.
Specifications
KIAA2013 | |
Polyclonal | |
Unconjugated | |
KIAA2013 | |
KIAA2013; RP5-1077B9.1; uncharacterized protein KIAA2013 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
90231 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IYS2 | |
KIAA2013 | |
Recombinant Human KIAA2013. Recombinant protein control fragment (Product #RP-107017). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction