Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIBRA Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282101
Description
KIBRA Polyclonal specifically detects KIBRA in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| KIBRA | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| WWC1 | |
| KIBRA antibody was raised against an 18 amino acid peptide near the amino terminus of human KIBRA. The immunogen is located within amino acids 60 - 110 of KIBRA. Amino Acid Squence: KTTQIEDPRVQWRREQEH | |
| Affinity Purified | |
| RUO | |
| 23286 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| FLJ10865, FLJ23369, HBeAg-binding protein 3, KIAA0869HBEBP3, KIBRAHBEBP36, Kidney and brain protein, protein KIBRA, protein WWC1, WW and C2 domain containing 1, WW domain-containing protein 1, WW, C2 and coiled-coil domain containing 1 | |
| Rabbit | |
| 110 kDa | |
| 0.1 mg | |
| Primary | |
| KIBRA antibody is human, mouse and rat reactive. At least three isoforms of KIBRA are known to exist; this antibody will detect all three isoforms. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction