Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIBRA Antibody (OTI1H8), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271732B
Description
KIBRA Monoclonal specifically detects KIBRA in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
KIBRA | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
WWC1 | |
Full length recombinant protein of human KIBRA (NP_056053) produced in HEK293T cells. | |
0.1 mL | |
23286.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1H8 | |
Western Blot | |
FLJ10865, FLJ23369, HBeAg-binding protein 3, KIAA0869HBEBP3, KIBRAHBEBP36, Kidney and brain protein, protein KIBRA, protein WWC1, WW and C2 domain containing 1, WW domain-containing protein 1, WW, C2 and coiled-coil domain containing 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction