Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIDINS220 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP150035V
Description
KIDINS220 Polyclonal antibody specifically detects KIDINS220 in Human samples. It is validated for Western BlotSpecifications
KIDINS220 | |
Polyclonal | |
Western Blot | |
kinase D-interacting substrate, 220kDa | |
The immunogen for this product maps to a region between residue 1721 and 1771 of human Ankyrin Repeat-Rich Membrane Spanning using the numbering given in entry NP_065789.1 (GeneID 57498). | |
0.1 mL | |
Phospho Specific | |
57498 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction