Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIDINS220 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100152
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100152 Supplier Invitrogen™ Supplier No. PA5100152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KIDINS220.

This gene encodes a transmembrane protein that is preferentially expressed in the nervous system where it controls neuronal cell survival, differentiation into exons and dendrites, and synaptic plasticity. The encoded protein interacts with membrane receptors, cytosolic signaling components, and cytoskeletal proteins, serving as a scaffold that mediates crosstalk between the neurotrophin pathway and several other intracellular signaling pathways. Aberrant expression of this gene is associated with the onset of various neuropsychiatric disorders and neurodegenerative diseases, including Alzheimer's disease. Naturally occurring mutations in this gene are associated with a syndrome characterized by spastic paraplegia, intellectual disability, nystagmus and obesity. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIDINS220
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Kidins220
Gene Accession No. Q9EQG6, Q9ULH0
Gene Alias 3110039L19Rik; AI194387; AI316525; ankyrin repeat-rich membrane spanning; ankyrin repeat-rich membrane spanning protein; ankyrin repeat-rich membrane-spanning protein; ARMS; C330002I19Rik; KIAA1250; KIDINS220; kinase D-interacting substance 220; kinase D-interacting substance of 220 kDa; kinase D-interacting substrate 220; kinase D-interacting substrate 220kDa; kinase D-interacting substrate of 220 kDa; kinase D-interacting substrate, 220kDa; MGC163482; mKIAA1250; truncated KIDINS220
Gene Symbols Kidins220
Host Species Rabbit
Immunogen A synthesized peptide derived from human KIDINS220(Accession Q9ULH0), corresponding to amino acid residues R381-T431.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116478, 57498, 77480
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.