Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Kidney Matched Tumor and Normal Tissue Lysate
SDP

Catalog No. NBP247100 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247100 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP247100 Supplier R&D Systems™ Supplier No. NBP247100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Two matched 1mg/mL vials (100μg/vial) of human kidney tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.

The Kidney Matched Tumor & Normal Tissue Lysate is derived from human.

Specifications

Quantity 2 Vials
Host Species Human
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.
Product Line Novus
Tissue Kidney
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.