Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Westcott™ Kids Scissors With Anti-Microbial Protection
SCI_ED

Catalog No. p-7214862
Click to view available options
Length (English):
5 in.
7 in.
Description:
Blunt
Pointed

Keeps products from degrading by preventing the growth of microorganisms. It's ideal for products that are shared around the home, office or school.

  • Handles are infused with anti-microbial protection to inhibit the growth of bacterial odors
  • Available in 4 bright colors (color can't be specified when ordered)
  • Stainless steel blades
  • Suitable for left or right-hand use
  • Blunt blades suitable for ages 4+
  • Ideal for cutting paper, creating craft projects and more
  • Manufacturer's limited lifetime warranty

Specifications

Product Type Scisssor
Color Assorted
Description Blunt
Grade Pre-K to 12
Age 4 to 18
Length (English) 5 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.