Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF1-binding protein Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297110PCP
Description
KIF1-binding protein Polyclonal antibody specifically detects KIF1-binding protein in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
KIF1-binding protein | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
DKFZP586B0923, KBP, KIAA1279, KIF1-binding protein, TTC20 | |
Produced in rabbits immunized with purified, recombinant Human KIF1-binding protein (Uniprot#: Q96EK5; Met1-Thr621) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
26128 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction