Learn More
Invitrogen™ KIF11 Polyclonal Antibody

Description
KIF11 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | KIF11 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | KIF11 |
| Gene Accession No. | P52732 |
| Gene Alias | Eg5; HKSP; KIF11; Kif8; Kifl1; Kines; kinesin 11; kinesin family member 11; kinesin-like 1; kinesin-like protein 1; Kinesin-like protein KIF11; kinesin-like spindle protein HKSP; Kinesin-related motor protein Eg5; Knsl1; MCLMR; Thyroid receptor-interacting protein 5; TR-interacting protein 5; TRIP5; TRIP-5 |
| Gene Symbols | KIF11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.