Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582394 Supplier Invitrogen™ Supplier No. PA582394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALIKEYTEEI ERLKRDLAAA REKNGVYISE ENFRVMSGKL TVQEEQIVEL IEKIGAVEEE LNRVTELFMD NKNELDQCKS DLQNKTQELE TTQKHLQETK LQLVK Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 83%.

This gene encodes a motor protein that belongs to the kinesin-like protein family. Members of this protein family are known to be involved in various kinds of spindle dynamics. The function of this gene product includes chromosome positioning, centrosome separation and establishing a bipolar spindle during cell mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIF11
Gene Accession No. P52732
Gene Alias Eg5; HKSP; KIF11; Kif8; Kifl1; Kines; kinesin 11; kinesin family member 11; kinesin-like 1; kinesin-like protein 1; Kinesin-like protein KIF11; kinesin-like spindle protein HKSP; Kinesin-related motor protein Eg5; Knsl1; MCLMR; Thyroid receptor-interacting protein 5; TR-interacting protein 5; TRIP5; TRIP-5
Gene Symbols KIF11
Host Species Rabbit
Immunogen Recombinant protein fragment of Human KIF11 (Product #RP-89481)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3832
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.