Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158181
Description
KIF12 Polyclonal specifically detects KIF12 in Human samples. It is validated for Western Blot.Specifications
KIF12 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
kinesin family member 12, kinesin-like protein KIF12, RP11-56P10.3 | |
Rabbit | |
Affinity purified | |
RUO | |
113220 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
B1ALC3 | |
KIF12 | |
Synthetic peptides corresponding to KIF12(kinesin family member 12) The peptide sequence was selected from the N terminal of KIF12. Peptide sequence SLGSPRPLPVRWNKTRGFYVEQLRVVEFGSLEALMELLQTGLSRRRNSAH. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Mouse: 92%; Chicken: 84%; Equine: 84%; Rat: 84%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction