Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554161
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554161 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554161 Supplier Invitrogen™ Supplier No. PA554161
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKCPRERSHS DWTQTRVLAE MLTEEEVVPS APPLPVRPPK TSPGLRGGAG VPNLAQRLEA LRDQIGSSLR RGRSQPPCSE GARSPGQVLP PH Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 61%.

KIF12 encodes a member of the kinesin superfamily of microtubule-associated molecular motors with functions related to the microtubule cytosekelton. Members of this superfamily play important roles in intracellular transport and cell division. A similar protein in mouse functions in the beta cell antioxidant signaling cascade, acting as a scaffold for the transcription factor specificity protein 1 (Sp1). Mice that lack this gene exhibit beta cell oxidative stress resulting in hypoinsulinemic glucose intolerance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF12
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIF12
Gene Accession No. Q96FN5
Gene Alias KIF12; kinesin 12; kinesin family member 12; kinesin superfamily protein 12; kinesin-like protein KIF12; N-9 kinesin; RP1156P10\.3
Gene Symbols KIF12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIF12. Recombinant protein control fragment (Product #RP-92968).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113220
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.