Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF13A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556096 Supplier Invitrogen™ Supplier No. PA556096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGLPLNLSNF VFCQYTFWDQ CESTVAAPVV DPEVPSPQSK DAQYTVTFSH CKDYVVNVTE EFLEFISDGA LAIEVWGHRC AGNGSSIWEV DSLHAK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.

This gene encodes a member of the kinesin family of microtubule-based motor proteins that function in the positioning of endosomes. This family member can direct mannose-6-phosphate receptor-containing vesicles from the trans-Golgi network to the plasma membrane, and it is necessary for the steady-state distribution of late endosomes/lysosomes. It is also required for the translocation of FYVE-CENT and TTC19 from the centrosome to the midbody during cytokinesis, and it plays a role in melanosome maturation. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF13A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIF13A
Gene Accession No. Q9H1H9
Gene Alias bA500C11.2; homolog of mouse KIF13A mannose-6-phosphate receptor transporter; KIF13A; kinesin family member 13A; kinesin-like protein KIF13A; Kinesin-like protein RBKIN; RBKIN
Gene Symbols KIF13A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIF13A. Recombinant protein control fragment (Product #RP-94857).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63971
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.