Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF13A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556097
Description
Immunogen sequence: ITSGYFSHSA SNATLSDMVV PSSDSSDQLA IQTKDADSTE HSTPSLVHDF RPSSNKELTE VEKGLVKDKI IVVPLKENSA LAKGSPSS Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 73%.
This gene encodes a member of the kinesin family of microtubule-based motor proteins that function in the positioning of endosomes. This family member can direct mannose-6-phosphate receptor-containing vesicles from the trans-Golgi network to the plasma membrane, and it is necessary for the steady-state distribution of late endosomes/lysosomes. It is also required for the translocation of FYVE-CENT and TTC19 from the centrosome to the midbody during cytokinesis, and it plays a role in melanosome maturation. Alternative splicing of this gene results in multiple transcript variants.
Specifications
| KIF13A | |
| Polyclonal | |
| Unconjugated | |
| KIF13A | |
| bA500C11.2; homolog of mouse KIF13A mannose-6-phosphate receptor transporter; KIF13A; kinesin family member 13A; kinesin-like protein KIF13A; Kinesin-like protein RBKIN; RBKIN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63971 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H1H9 | |
| KIF13A | |
| Recombinant protein corresponding to Human KIF13A. Recombinant protein control fragment (Product #RP-94856). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction