Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF13A Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501559194
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9194 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9194 Supplier Bethyl Laboratories, Inc Supplier No. A301078AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Plus end-directed microtubule-dependent motor protein involved in mannose-6-phosphate receptor (M6PR) transport to the plasma membrane.

Specifications

Antigen KIF13A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene KIF13A
Gene Accession No. Q9H1H9
Gene Alias RBKIN, bA500C11.2, homolog of mouse KIF13A mannose-6-phosphate receptor transporter, kinesin-like protein KIF13A, kinesin-like protein RBKIN
Gene Symbols KIF13A
Host Species Rabbit
Immunogen Between 1725 and 1775
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63971
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.