Learn More
KIF13A Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301078A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Plus end-directed microtubule-dependent motor protein involved in mannose-6-phosphate receptor (M6PR) transport to the plasma membrane.Specifications
| KIF13A | |
| Polyclonal | |
| Unconjugated | |
| KIF13A | |
| RBKIN, bA500C11.2, homolog of mouse KIF13A mannose-6-phosphate receptor transporter, kinesin-like protein KIF13A, kinesin-like protein RBKIN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63971 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9H1H9 | |
| KIF13A | |
| Between 1725 and 1775 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.