Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF13A Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502207540
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7540 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7540 Supplier Fabgennix Inc Supplier No. KIF13A101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIF13A Polyclonal antibody specifically detects KIF13A in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen KIF13A
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene KIF13A
Gene Accession No. Q9EQW7, Q9H1H9
Gene Alias bA500C11.2; homolog of mouse KIF13A mannose-6-phosphate receptor transporter; KIF13A; kinesin family member 13A; kinesin-like protein KIF13A; Kinesin-like protein RBKIN; RBKIN
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1700-1800 on human KIF13A isoform a protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16553, 308173, 63971
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.