Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF13B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540807
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540807 Supplier Invitrogen™ Supplier No. PA540807
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SGKSYTMMGT ADQPGLIPRL CSGLFERTQK EENEEQSFKV EVSYMEIYNE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

KIF13B (Kinesin Family Member 13B) is a Protein Coding gene. Diseases associated with KIF13B includeSpastic Paraplegia 37, Autosomal DominantandSpastic Paraplegia 11, Autosomal Recessive. Among its related pathways areGolgi-to-ER retrograde transportandFactors involved in megakaryocyte development and platelet production. Gene Ontology (GO) annotations related to this gene includeprotein kinase bindingandmicrotubule motor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF13B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KIF13B
Gene Accession No. Q9NQT8
Gene Alias 5330429L19Rik; 6030414C01; AI429803; C130021D12Rik; GAKIN; guanylate kinase associated kinesin; KIAA0639; Kif13b; kinesin 13B; kinesin family member 13B; kinesin-like protein GAKIN; Kinesin-like protein KIF13B; N-3 kinesin
Gene Symbols KIF13B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KIF13B (aa 107-156).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23303
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.