Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF13B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599645
Description
Antibody detects endogenous levels of total KIF13B.
KIF13B (Kinesin Family Member 13B) is a Protein Coding gene. Diseases associated with KIF13B includeSpastic Paraplegia 37, Autosomal DominantandSpastic Paraplegia 11, Autosomal Recessive. Among its related pathways areGolgi-to-ER retrograde transportandFactors involved in megakaryocyte development and platelet production. Gene Ontology (GO) annotations related to this gene includeprotein kinase bindingandmicrotubule motor activity.
Specifications
KIF13B | |
Polyclonal | |
Unconjugated | |
KIF13B | |
5330429L19Rik; 6030414C01; AI429803; C130021D12Rik; GAKIN; guanylate kinase associated kinesin; KIAA0639; Kif13b; kinesin 13B; kinesin family member 13B; kinesin-like protein GAKIN; Kinesin-like protein KIF13B; N-3 kinesin | |
Rabbit | |
Affinity chromatography | |
RUO | |
16554, 23303 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NQT8 | |
KIF13B | |
A synthesized peptide derived from human KIF13B(Accession Q9NQT8), corresponding to amino acid residues S1644-A1694. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction