Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF14 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100254AF532
Description
KIF14 Polyclonal antibody specifically detects KIF14 in Human samples. It is validated for Western BlotSpecifications
KIF14 | |
Polyclonal | |
Western Blot | |
KIAA0042MGC142302, kinesin family member 14, kinesin-like protein KIF14 | |
The immunogen recognized by this antibody maps to a region between residues 900 and 950 of human Kinesin Family Member 14 using the numbering given in entry NP_055690.1 (GeneID 9928). | |
0.1 mL | |
Cell Biology, Cellular Markers, DNA Repair | |
9928 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction