Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584368
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584368 Supplier Invitrogen™ Supplier No. PA584368
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKLSMMIQEA NAISSKLKTY YVFGRHDISD KSSSDTSIRV RNLKLGISTF WSLEKFESKL AAMKELYESN GSNRGEDA Highest antigen sequence indentity to the following orthologs: Rat - 59%, Mouse - 81%.

This gene encodes a member of the kinesin-3 superfamily of microtubule motor proteins. These proteins are involved in numerous processes including vesicle transport, chromosome segregation, mitotic spindle formation, and cytokinesis. In human HeLa-S3 and 293T cells, this protein is localized to the cytoplasm during interphase, to the spindle poles and spindle microtubules during mitosis, and to the midbody during cytokinesis. An internal motor domain displays microtubule-dependent ATPase activity, consistent with its function as a microtubule motor protein. Knockdown of this gene results in failed cytokinesis with endoreplication, which results in multinucleated cells. This gene has been identified as a likely oncogene in breast, lung and ovarian cancers, as well as retinoblastomas and gliomas. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF14
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIF14
Gene Accession No. Q15058
Gene Alias D1Ertd367e; E130203M01; KIAA0042; KIF14; kinesin family member 14; kinesin-like protein KIF14; MKS12; N-3 kinesin; truncated kinesin family member 14
Gene Symbols KIF14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIF14. Recombinant protein control fragment (Product #RP-106832).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9928
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.