Learn More
KIF15 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302706AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Plus-end directed kinesin-like motor enzyme involved in mitotic spindle assembly.Specifications
KIF15 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9NS87 | |
KIF15 | |
Between 1225 and 1275 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
KIF15 | |
HKLP2, kinesin-like 7, kinesin-like protein 2, kinesin-like protein 7, KNSL7, NY-BR-62, serologically defined breast cancer antigen NY-BR-62 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56992 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.