Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF16B Polyclonal Antibody
KIF16B Polyclonal Antibody
Supplier: Thermo Scientific PA562343
Description
KIF16B Polyclonal Antibody for IHC

Specifications
KIF16B | |
Polyclonal | |
Unconjugated | |
KIF16B | |
C20orf23; KIAA1590; KIF16B; kinesin family member 16B; kinesin motor protein; kinesin-like protein KIF16B; KISC20ORF; SNX23; sorting nexin 23; Sorting nexin-23; testis secretory sperm-binding protein Li 201a | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55614 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96L93 | |
KIF16B | |
Recombinant protein corresponding to Human KIF16B. Recombinant protein control fragment (Product #RP-99315). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction