Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF17 Polyclonal Antibody

Catalog No. PA125040
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA125040 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA125040 Supplier Invitrogen™ Supplier No. PA125040

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain cytosolic fraction, PC12, NGF.

Dendrite-specific motor protein which, in association with the Apba1-containing complex (LIN-10-LIN-2-LIN-7 complex), transports vesicles containing N-methyl-D-aspartate (NMDA) receptor subunit NR2B along microtubules. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF17
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene Kif17
Gene Accession No. Q99PW8
Gene Alias 5930435E01Rik; AW492270; KIAA1405; Kif17; KIF17 variant protein; Kif17b; KIF3-related motor protein; KIF3X; kinesin 17; kinesin family member 17; kinesin-like protein KIF17; KLP-2; mKIAA1405; MmKIF17; N-4 kinesin; OSM-3; RGD1562511
Gene Symbols Kif17
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 589-606 of mouse KIF17 conjugated to KLH.
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16559, 500571
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.