Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530778 Supplier Invitrogen™ Supplier No. PA530778
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Human (99%), Mouse (88%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Dendrite-specific motor protein which, in association with the Apba1-containing complex (LIN-10-LIN-2-LIN-7 complex), transports vesicles containing N-methyl-D-aspartate (NMDA) receptor subunit NR2B along microtubules. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Kif17
Gene Accession No. Q9P2E2
Gene Alias 5930435E01Rik; AW492270; KIAA1405; Kif17; KIF17 variant protein; Kif17b; KIF3-related motor protein; KIF3X; kinesin 17; kinesin family member 17; kinesin-like protein KIF17; KLP-2; mKIAA1405; MmKIF17; N-4 kinesin; OSM-3; RGD1562511
Gene Symbols Kif17
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human KIF17. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57576
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.