Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109878 Supplier Invitrogen™ Supplier No. PA5109878
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKAILTQQMS PSSLSALLSR QVPPDPVQVE EKLLPQPVIQ HDVEAEKQLI REEYEERLAR LKADYKAEQE SRARLEEDIT AMRNSYDVRL STLEENLRKE TEAVLQVGVL YKAEVMSRAE FASSAEYPPA FQYETVVKPK VFSTTDTLPS DDVSKTQVSS RFAELPKVEP SKSEISLGSS ESSSLEETSV SEAFPGPEEP SNVEVSMPTE ESRSRYFLDE CLGQEAAGHL LGEQNYLPQE EPQEVPLQGL LGLQDPFAEV EAKLARLSST VARTDAPQAD VPKVPVQVPA PTDLLEPSDA R.

Dendrite-specific motor protein which, in association with the Apba1-containing complex (LIN-10-LIN-2-LIN-7 complex), transports vesicles containing N-methyl-D-aspartate (NMDA) receptor subunit NR2B along microtubules. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF17
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Kif17
Gene Accession No. Q99PW8, Q9P2E2
Gene Alias 5930435E01Rik; AW492270; KIAA1405; Kif17; KIF17 variant protein; Kif17b; KIF3-related motor protein; KIF3X; kinesin 17; kinesin family member 17; kinesin-like protein KIF17; KLP-2; mKIAA1405; MmKIF17; N-4 kinesin; OSM-3; RGD1562511
Gene Symbols Kif17
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16559, 500571, 57576
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.