Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115468 Supplier Thermo Scientific Supplier No. PA5115468
Add to Cart
Add to Cart

KIF17 Polyclonal Antibody

KIF17 Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF17
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Kif17
Gene Accession No. Q99PW8, Q9P2E2
Gene Alias 5930435E01Rik; AW492270; KIAA1405; Kif17; KIF17 variant protein; Kif17b; KIF3-related motor protein; KIF3X; kinesin 17; kinesin family member 17; kinesin-like protein KIF17; KLP-2; mKIAA1405; MmKIF17; N-4 kinesin; OSM-3; RGD1562511
Gene Symbols Kif17
Host Species Rabbit
Immunogen A synthesized peptide derived from human KIF17(Accession Q9P2E2), corresponding to amino acid residues L861-T911.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16559, 500571, 57576
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.