Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF18A Antibody, Novus Biologicals™
SDP

Catalog No. p-200037836 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB432362 25ul
NBP185126 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB432362 Supplier Novus Biologicals Supplier No. NBP18512625UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

KIF18A Polyclonal specifically detects KIF18A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen KIF18A
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q8NI77
Gene Alias DKFZP434G2226, kinesin family member 18A, kinesin-like protein KIF18A, Marrow stromal KIF18A, MS-KIF18A
Gene Symbols KIF18A
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:LHCHHLHLQNKDLKAQIRHMMDLACLQEQQHRQTEAVLNALLPTLRKQYCTLKEAGLSNAAFESDFKEIEHLVERKKVVVWADQTAEQPKQND
Molecular Weight of Antigen 102 kDa
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81930
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.