Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF18A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558808
Description
Immunogen sequence: AFQNPSTVTL MKPSSFTTSF QAISSNINSD NCLKMLCEVA IPHNRRKECG QEDLDSTFTI CEDIKSSKCK LPEQESLPND NKDILQRLDP Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 33%.
May have a role in a signaling pathway. Could act as a tumor suppressor.
Specifications
KIF18A | |
Polyclonal | |
Unconjugated | |
KIF18A | |
AU024633; B130001M12Rik; Kif18a; kinesin family member 18A; kinesin superfamily protein 18A; kinesin-like protein KIF18A; Marrow stromal KIF18A; MS-KIF18A; N-8 kinesin; OK/SW-cl.108; PPP1R99; protein phosphatase 1, regulatory subunit 99 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81930 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NI77 | |
KIF18A | |
Recombinant protein corresponding to Human KIF18A. Recombinant protein control fragment (Product #RP-97336). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction