Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF18A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106359
Description
Antibody detects endogenous levels of total KIF18A.
May have a role in a signaling pathway. Could act as a tumor suppressor.
Specifications
KIF18A | |
Polyclonal | |
Unconjugated | |
KIF18A | |
AU024633; B130001M12Rik; Kif18a; kinesin family member 18A; kinesin superfamily protein 18A; kinesin-like protein KIF18A; Marrow stromal KIF18A; MS-KIF18A; N-8 kinesin; OK/SW-cl.108; PPP1R99; protein phosphatase 1, regulatory subunit 99 | |
Rabbit | |
Affinity chromatography | |
RUO | |
228421, 81930 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8NI77, Q91WD7 | |
KIF18A | |
A synthesized peptide derived from human KIF18A(Accession Q8NI77), corresponding to amino acid residues Q216-S266. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction