Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF18A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531477
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531477 Supplier Invitrogen™ Supplier No. PA531477
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HCT116. Predicted reactivity: Mouse (88%), Chicken (80%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May have a role in a signaling pathway. Could act as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF18A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene KIF18A
Gene Accession No. Q8NI77
Gene Alias AU024633; B130001M12Rik; Kif18a; kinesin family member 18A; kinesin superfamily protein 18A; kinesin-like protein KIF18A; Marrow stromal KIF18A; MS-KIF18A; N-8 kinesin; OK/SW-cl.108; PPP1R99; protein phosphatase 1, regulatory subunit 99
Gene Symbols KIF18A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human KIF18A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81930
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.